Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4000

Catalog No. p-7052084
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42500AD 50 mL
AA42500AK 250 mL
AA42500K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42500AD Supplier Thermo Scientific Chemicals Supplier No. 042500AD
Only null left
Add to Cart

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Viscous Liquid
Boiling Point >225°C
Melting Point <-40°C
Density 0.960 g/mL
Solubility Information Slightly miscible with water. Immiscible with methanol and ethylene glycol.
Quantity 50 mL
Flash Point 285°C (545°F)
Odor Odorless
Refractive Index 1.4015
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 4,000
Show More Show Less
RTECSNumber TQ2690000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.