Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polydisperse Particle Standard, Particle Size Standard, PS224, MilliporeSigma™ Supelco™

Catalog No. 111002590 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 0.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2590 10 x 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2590 Supplier MilliporeSigma Supelco Supplier No. 5756310X0.5G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 1313-59-3
Physical Form Particles
Type Certified Reference Material
Quantity 10 x 0.5 g
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.