Learn More
Sigma Aldrich Fine Chemicals Biosciences PolyDL lactide co glycolid5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P19415G
Encompass
The product is an amorphous solid with a transition temperature (Tg) in the range of 45-50C. It is soluble in most organic solvents such as methylene chloride tetrahydrofuran ethyl acetate acetone chloroform and hexafluoroispropanol.
Catalog No. 50-290-4186