Learn More
MSC Rubbermaid Lid, Polyethylene, Square, 12-22 qt
Catalog No. 502837241
Encompass
This polyethylene lid is designed for use with space-saving square containers, providing a secure and leak-resistant seal. It features a corner-pull tab for easy removal and is suitable for various food storage applications, meeting multiple food-grade standards.
  • Corner-pull tab for easy removal
  • Snap-tight lid for a secure fit
  • Leak-resistant design
  • Made from food-grade polyethylene
  • Compatible with 12 qt, 18 qt, and 22 qt square containers
  • Withstands temperatures from -20°F to 150°F
  • NSF, REACH, USDA Approved, and TUV Rheinland Food-Grade certified
Catalog No. 50-283-7241 Supplier MSC Supplier No. 43837012
This polyethylene lid is designed for use with space-saving square containers, providing a secure and leak-resistant seal. It features a corner-pull tab for easy removal and is suitable for various food storage applications, meeting multiple food-grade standards.
  • Corner-pull tab for easy removal
  • Snap-tight lid for a secure fit
  • Leak-resistant design
  • Made from food-grade polyethylene
  • Compatible with 12 qt, 18 qt, and 22 qt square containers
  • Withstands temperatures from -20°F to 150°F
  • NSF, REACH, USDA Approved, and TUV Rheinland Food-Grade certified