Learn More
Grainger Hand Operated Drum Pump, Piston, Basic Pump with Discharge Hose, For Container Type Drum
Catalog No. NC1332066
Encompass_Preferred
Diesel exhaust fluid (DEF) hand drum pumps are used specifically with DEF to maintain purity when filling the reservoir of a diesel engine's Selective Catalytic Reduction (SCR) system. These pumps are certified to ISO standards that regulate the handling, transportation, and storage of DEF. They are used for dispensing and transferring small volumes of DEF when servicing late model trucks and SUVs as well as SCR-based stationary industrial systems that operate on diesel fuel. Hand drum pumps are manually powered devices operated by a crank, lever, or other mechanism to draw fluids out of a container. They allow pumping where there's no access to a power source and are ideal for dispensing and transferring small amounts of fluid. Sometimes called a manual or rotary drum pump, these hand-operated pumps connect to an adapter that allows the shaft of the pump to be mounted through the bung hole of a drum or other container.
Catalog No. NC1332066 Supplier Grainger Supplier No. 4HA27
Diesel exhaust fluid (DEF) hand drum pumps are used specifically with DEF to maintain purity when filling the reservoir of a diesel engine's Selective Catalytic Reduction (SCR) system. These pumps are certified to ISO standards that regulate the handling, transportation, and storage of DEF. They are used for dispensing and transferring small volumes of DEF when servicing late model trucks and SUVs as well as SCR-based stationary industrial systems that operate on diesel fuel. Hand drum pumps are manually powered devices operated by a crank, lever, or other mechanism to draw fluids out of a container. They allow pumping where there's no access to a power source and are ideal for dispensing and transferring small amounts of fluid. Sometimes called a manual or rotary drum pump, these hand-operated pumps connect to an adapter that allows the shaft of the pump to be mounted through the bung hole of a drum or other container.