Learn More
Sigma Aldrich Polyester Bis-mpa Dendron 2 Carboxyl 1 Biotin (core) Generation 1
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Polymer architecture: Dendron
- End Group Functionality: Boc-protected amine
- Boc: tert-Butyloxycarbonyl
- Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid. Bis-MPA (or 2,2-Bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers.
- Dendrons synthesized from bis-MPA are biodegradable and exhibit low cytotoxicity, making them well-suited for use in biological research applications. These materials have been utilized in various applications, including signal amplification in bioassays and drug delivery. After Boc deprotection, amine-functionalized dendrons can be readily used in EDC or DCC coupling reactions with carbonyl-containing compounds to yield highly functionalized materials for a variety of biomedical applications. Additionally, the azide-core can be readily conjugated using either copper(I)-catalyzed alkyne-azide cycloaddition (CuAAC) or strain-promoted alkyne-azide cycloaddition (SPAAC).
Specifications
Specifications
| Molecular Formula | C31H48N2O16S |
| Linear Formula | C31H48N2O16S |
| Quantity | 50 mg |
| Synonym | Bis-MPA dendron; PFd-G1-Biotin-COOH |
| Molecular Weight (g/mol) | 736.78 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.