Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyester bis-MPA dendron 2 NHBoc, 1 azide (core)

Catalog No. 901311100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-131-1100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-131-1100MG Supplier Sigma Aldrich Supplier No. 901311100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Polymer architecture: Dendron
  • End Group Functionality: Boc-protected amine
  • Boc: tert-Butyloxycarbonyl
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid. Bis-MPA (or 2,2-Bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers.
  • Dendrons synthesized from bis-MPA are biodegradable and exhibit low cytotoxicity, making them well-suited for use in biological research applications. These materials have been utilized in various applications, including signal amplification in bioassays and drug delivery. After Boc deprotection, amine-functionalized dendrons can be readily used in EDC or DCC coupling reactions with carbonyl-containing compounds to yield highly functionalized materials for a variety of biomedical applications. Additionally, the azide-core can be readily conjugated using either copper(I)-catalyzed alkyne-azide cycloaddition (CuAAC) or strain-promoted alkyne-azide cycloaddition (SPAAC).

Specifications

Molecular Formula C27H47N5O10
Linear Formula C27H47N5O10
Quantity 100 mg
Synonym Bis-MPA dendron; PFd-G1-Azide-NH-BOC
Molecular Weight (g/mol) 601.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.