Learn More
MSC Value Collection Polyester Solid Braid Rope, 7/32 in. Diameter, 200 ft. Length, 1 ea.
Catalog No. 5000514453
Encompass
This polyester solid braid rope is designed for general utility applications requiring reliable performance. The solid braid construction offers excellent flexibility and durability for various industrial and maintenance tasks.
  • Constructed from durable polyester material
  • Features a solid braid design for enhanced strength and flexibility
  • Measures 7/32 in. in diameter
  • Provides a working load limit of 118 lb.
  • Supplied in a 200 ft. coil
Catalog No. 50-005-14453 Supplier MSC Supplier No. 45901071
This polyester solid braid rope is designed for general utility applications requiring reliable performance. The solid braid construction offers excellent flexibility and durability for various industrial and maintenance tasks.
  • Constructed from durable polyester material
  • Features a solid braid design for enhanced strength and flexibility
  • Measures 7/32 in. in diameter
  • Provides a working load limit of 118 lb.
  • Supplied in a 200 ft. coil