Learn More
Brady™ Polyester Caution Sign: FLAMMABLE
Catalog No. 19107392 Shop All Brady Corporation Products
Change view
Click to view available options
Adhesive Type:
Permanent Acrylic
Catalog No. Adhesive Type
19-107-392 Permanent Acrylic
1 options
Catalog No. 19-107-392 Supplier Brady™ Supplier No. 80357
Add to Cart
Add to Cart
Improve workplace safety with proper signage. The Brady™ Caution Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
  • Polyester material is
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Mounting Type Self-adhesive
Displayed Text FLAMMABLE
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.