Learn More
ACL Inc Staticide 6" Polester Knit swabs with elongated tip. 100 swabs per bag.
Catalog No. NC1841789
Encompass_Preferred
DOUBLE LAYER Polyester Knit Non-Abrasive Lint Free, Highly absorbent. Free of silicone, amides,and DOP contamination, elongated tip, ISO Class 4 - 5, Class 10 laundered and packaged.
Catalog No. NC1841789 Supplier ACL Inc Supplier No. 7060
DOUBLE LAYER Polyester Knit Non-Abrasive Lint Free, Highly absorbent. Free of silicone, amides,and DOP contamination, elongated tip, ISO Class 4 - 5, Class 10 laundered and packaged.