Learn More
ACL Inc Staticide 8399PK Polyester Knit Wipes - 9” x 9”; 150 wipes per bag; 8 bags per case
Catalog No. NC1841774
Encompass_Preferred
High Strength and Non-Linting
ACL Staticide 8399PK Polyester Knit Wipes are an economical choice for those seeking an all-purpose, lint-free wipe. This lightweight, double knit wipe is made in an interlocking pattern with continuous filament polyester yarn for high strength and abrasion resistance even on rough surfaces. The soft texture of the wipe protects surfaces from scratching even when force is applied.
Ideal Applications:
Special applications needing no lint or fiber loss
Critical environment and process cleaning
Product Features:
Suitable for ISO Class 5 - 7 (Class 100 - 10,000) environments
100% pure polyester
Ultra-low particle and fiber generation
Super soft material; does not scratch
Laser-sealed edges
Free of silicon, amides, and DOP contamination
Class 100 laundered and packaged
Catalog No. NC1841774 Supplier ACL Inc Supplier No. 8399PK
High Strength and Non-Linting
ACL Staticide 8399PK Polyester Knit Wipes are an economical choice for those seeking an all-purpose, lint-free wipe. This lightweight, double knit wipe is made in an interlocking pattern with continuous filament polyester yarn for high strength and abrasion resistance even on rough surfaces. The soft texture of the wipe protects surfaces from scratching even when force is applied.
Ideal Applications:
Special applications needing no lint or fiber loss
Critical environment and process cleaning
Product Features:
Suitable for ISO Class 5 - 7 (Class 100 - 10,000) environments
100% pure polyester
Ultra-low particle and fiber generation
Super soft material; does not scratch
Laser-sealed edges
Free of silicon, amides, and DOP contamination
Class 100 laundered and packaged