Learn More
ELKO FILTERING CO POLYESTER MESH 40 MICR 40"X1YD
SDP
Supplier:  ELKO FILTERING CO 0740/25
Encompass
Polyester Mesh 40 Micron - Open Area %: 25 - Width: 40 in (sold by the yard)
Catalog No. NC1464953