Learn More
ELKO FILTERING CO POLYESTER MESH 500UM 1 YARD
SDP
Supplier:  ELKO FILTERING CO 07500/48
Encompass
Nylon Monofilament Precision Woven Mesh - 500 Micron Opening, Open Area 48%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 07-500/48
Catalog No. NC0523929