Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, Transparent, 0.4 micron, 12 Micron Thickness, 2E6 pores/cm2, 25mm, 100/pack
SDP
Catalog No. NC1721128
Encompass_Preferred
Polyester (PETE) Membrane Filters, Transparent, 0.4 micron, 12 Micron Thickness, 2E6 pores/cm2, 25mm, 100/pack
Catalog No. NC1721128 Supplier Sterlitech Corporation Supplier No. 1300016
Polyester (PETE) Membrane Filters, Transparent, 0.4 micron, 12 Micron Thickness, 2E6 pores/cm2, 25mm, 100/pack