Learn More
Andwin Scientific POLYESTER PLATE FILM, STERILE
SDP
Catalog No. NC0605802
Encompass_Preferred
The sterile polyester plate film for ELISA and General Incubation allows a reliable seal on every well. It minimizes evaporation, prevents spillage, contamination and edge effects. It can also be used with tissue culture plates.
Catalog No. NC0605802 Supplier Andwin Scientific Supplier No. 60941064
The sterile polyester plate film for ELISA and General Incubation allows a reliable seal on every well. It minimizes evaporation, prevents spillage, contamination and edge effects. It can also be used with tissue culture plates.