Learn More
Brady™ Polyester Sign: DANGER - ELECTRICAL ROOM. NO STORAGE PERMITTED.
Catalog No. 19109502 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Polyester
Catalog No. Material
19-109-502 Polyester
1 options
Catalog No. 19-109-502 Supplier Brady™ Supplier No. 127004
  • $22.25
    $15.65 / Each of 1
    Save $6.60
    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Polyester
Shape Rectangular
Languages English
Dimensions (L x W) 10 x 7 in.
Displayed Text DANGER - ELECTRICAL ROOM. NO STORAGE PERMITTED.
Product Type Safety Sign
Color Red on White
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.