Learn More
Brady™ Polyester Sign: NOTICE - MATERIAL SAFETY DATA SHEETS AVAILABLE UPON REQUEST (English/Spanish)
Shop All Brady Corporation ProductsDescription
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Specifications
| Material | Polyester |
| Shape | Rectangular |
| Dimensions (L x W) | 14 x 10 in. |
| Displayed Text | NOTICE - MATERIAL SAFETY DATA SHEETS AVAILABLE UPON REQUEST (English/Spanish) |
| Color | Blue on White |