Learn More
Valutek™ Polyester Spunlace Non-woven Wiper, Cold Knife Cut
Catalog No. 191502018
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
18 x 18 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-150-2018 12 x 12 in.
19-150-2019 18 x 18 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-2018 Supplier Valutek™ Supplier No. VTSNTR1212
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic yarn, which has a low level of particulate and extractable counts.
  • Valutek™’s spunlace non-woven wiper is constructed from a hydro entangled, nonwoven blend of 55% cellulose and 45% polyester
  • Cold knife cut edge Contains no chemical binders
  • High absorbency and low abrasion resistance
  • Chemically compatible with common cleaning and disinfecting solutions
  • Low levels of extractable and moderate particle counts
  • Available in a variety of sizes to suit most applications
  • All Valutek™ wipers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
  • This product is recommended for use in a cleanroom Class 10,000+ (ISO 7+) critical environment
  • The spunlace non-woven wiper is commonly called the “multi-tasking” or “grunt” wipe. It is a highly absorbent wiper for general purpose and environmental / facility cleaning. It is not suitable for direct product contact
Specifications
Material Polyester
Dimensions (L x W) 12 x 12 in.
Sterility Non-sterile