Learn More
Electron Microscopy Sciences Knit Sealed Polyester All-Purpose Swabs, 1.6 mm tips
SDP
Catalog No. 501930635
Encompass_Preferred
Very clean and durable all-purpose swabs are acid and solvent resistant. Sealed edges build in high strength with no loose fibers or particulates. They are ideal for tough scrubbing around raised shapes. Cleanroom process results in the lowest ionic, non-volatile residue and particle contaminants. Polypropylene handle.
Tips: 1.6 mm
Catalog No. 50-193-0635 Supplier Electron Microscopy Sciences Supplier No. 7832580
Very clean and durable all-purpose swabs are acid and solvent resistant. Sealed edges build in high strength with no loose fibers or particulates. They are ideal for tough scrubbing around raised shapes. Cleanroom process results in the lowest ionic, non-volatile residue and particle contaminants. Polypropylene handle.
Tips: 1.6 mm