Learn More
Brady™ Polyester Warning Sign: ACID
Catalog No. 19107393 Shop All Brady Corporation Products
Catalog No. 19-107-393 Supplier Brady™ Supplier No. 80358
Improve workplace safety with proper signage. The Brady™ Warning Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
  • Polyester material is
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Acrylic
Mounting Type Self-adhesive
Displayed Text ACID