Learn More
Electron Microscopy Sciences Polyester Wax 500 GR
SDP
Catalog No. 5027951
Encompass_Preferred
A synthetic polyester wax with a very low melting point (37 degree C) used as a ribboning embedding medium. Its main advantage over paraffin and ester waxes is its low melting point, which reduces tissue hardening, shrinkage, and eliminates the need for infiltration ovens.
The wax is soluble in most organic solvents, including alcohol, ethers, esters, ketones and hydrocarbons. The wax is water tolerant, almost opaque, and sections easily. No electrification of ribbons occurs during sectioning. Sections of 2 microns and more may be cut at room temperatures between 10 and 22 degree C. It is advantageous to keep the wax in its solid state prior to use.
Catalog No. 50-279-51 Supplier Electron Microscopy Sciences Supplier No. 19312
Add to Cart
Edge
Add to Cart
A synthetic polyester wax with a very low melting point (37 degree C) used as a ribboning embedding medium. Its main advantage over paraffin and ester waxes is its low melting point, which reduces tissue hardening, shrinkage, and eliminates the need for infiltration ovens.
The wax is soluble in most organic solvents, including alcohol, ethers, esters, ketones and hydrocarbons. The wax is water tolerant, almost opaque, and sections easily. No electrification of ribbons occurs during sectioning. Sections of 2 microns and more may be cut at room temperatures between 10 and 22 degree C. It is advantageous to keep the wax in its solid state prior to use.