Learn More
TrippNT POLYETHENE MEDICAL MRI CORE DX
SDP
Supplier:  TrippNT 51728
Encompass_Preferred
TrippNT's MRI Core DX with White Drawers and Sliding Door TrippNT's is compatible for use within the sensitive magnetic imaging environment, keeping supplies mobile and organized. The cutaway ergonomic grab handles in the top make the Core DX easy to maneuver. Constructed from non-ferrous durable high density polyethylene and ABS and non-ferrous casters and screws. Ships fully Assembled.
Catalog No. NC3720657