Learn More
Cayman Chemical Polyethylen Glycol RabbIt Mo
SDP
Catalog No. 502947813
Encompass_Preferred
Immunogen: KLH-PEG with terminal methoxy group • Host: Rabbit • Species Reactivity: Species independent • Cross Reactivity: (+) Methoxy-PEG • Applications: ELISA, IHC, WB
Catalog No. 50-294-7813 Supplier Cayman Chemical Supplier No. 32180100 UG
Immunogen: KLH-PEG with terminal methoxy group • Host: Rabbit • Species Reactivity: Species independent • Cross Reactivity: (+) Methoxy-PEG • Applications: ELISA, IHC, WB