Learn More
Braintree Scientific Polyethylene .023"""" x .038"""" Per Ft., 500 ft. Continuous
SDP
Catalog No. 501955512
Encompass
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**
Catalog No. 50-195-5512 Supplier Braintree Scientific Supplier No. PE50500FT
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**