Learn More
Braintree Scientific Polyethylene .030"""" x .048"""" Per Ft., 500 ft. Continuous
SDP
Catalog No. 501955517
Encompass
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**
Catalog No. 50-195-5517 Supplier Braintree Scientific Supplier No. PE60500FT
Polyethylene Tubing Polyethylene is a relatively rigid polymer suitable for short-term catheterizations and for ex vivo applications. Polyethylene sleeves can be slipped over less rigid catheter materials (MicroRenathane, RenaSil™) at suture points to protect the latter from inadvertent constriction **Sold for research purposes only and not for human use**