Learn More
VGuard™ Polyethylene Arm Sleeve
Catalog No. 17005969
Click to view available options
Color:
Blue
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-005-969 White
17-005-970 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-005-969 Supplier VGuard™ Supplier No. B76N10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight and water repellent arm protection.
  • Disposable
  • Flexible polyethylene for all-day wear
  • Keeps skin and hair from contaminating product
  • Protects against dust, dirt, grime, and fluids
  • Blue color style aids in contamination control
Specifications
Product Type General Purpose Sleeve
Material Polyethylene
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Length (Metric) 457.2 mm