Learn More
Research Products International Corp Nalgene Polyethylene Bottle with Screw Cap, 1 Gallon Capacity
SDP
Catalog No. 50213081
Encompass
Store chemicals and standards and collect and store distilled water. Nalgene Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols.
Can be used in freezer to -100°C (-148°F) for long term storage and features a built-in shoulder loop for attaching identification tag. Translucence allows for easy viewing of liquid levels and linerless caps provide the ultimate in leakproof protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents.
Catalog No. 50-213-081 Supplier Research Products International Corp Supplier No. 22020010
Store chemicals and standards and collect and store distilled water. Nalgene Large Narrow-Mouth LDPE Bottles are translucent and offer flexibility, impact resistance and visibility of contents. The extremely low trace metal content in low density polyethylene makes them ideal for trace metal analysis. These bottles provide excellent chemical resistance to most acids, bases and alcohols.
Can be used in freezer to -100°C (-148°F) for long term storage and features a built-in shoulder loop for attaching identification tag. Translucence allows for easy viewing of liquid levels and linerless caps provide the ultimate in leakproof protection without the use of liner that can wrinkle, cause leaks or contaminate your reagents.