Learn More
Fisherbrand™ Polyethylene Cleanroom Clipboards
Catalog No. 18003109 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
9 x 12 in.
9 x 14.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
18-003-109 9 x 12 in.
18-003-110 9 x 14.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-003-109
Add to Cart
Add to Cart
Cleanroom clipboard/ ESD-safe for cleanroom documentation.
  • Cleanroom clipboards and clips are made from polyethylene to reduce the possibility of ESDs and particle contamination.
  • Boards can be easily wiped down with IPA and colors will not fade with use and time.
  • This product is ideal for Class 10 to Class 1000 cleanroom environments.
Specifications
Material Polyethylene
Color White
Dimensions (L x W) 9 x 12 in.
Product Type Cleanroom Clipboard