Learn More
Electron Microscopy Sciences Nalgene Polyethylene Dewar Flask 4L, 324 mm height, 146 mm I.D.
SDP
Catalog No. 501899064
Encompass_Preferred
High density polyethylene Dewar Flask with vented and insulated HDPE covers. They are unbreakable, chemically resistant, and withstand temperatures from -196 to 100 degree C) (-321 to +212 degree F). Double walls are filled with foamed urethane to minimize heat exchange. May be used for short term storage of dry ice-solvent, liquid nitrogen, or for use as a hot bath. They are ideal for storing frozen biological specimens.
Height: 324 mm
I.D.: 146 mm
Catalog No. 50-189-9064 Supplier Electron Microscopy Sciences Supplier No. 6203804
High density polyethylene Dewar Flask with vented and insulated HDPE covers. They are unbreakable, chemically resistant, and withstand temperatures from -196 to 100 degree C) (-321 to +212 degree F). Double walls are filled with foamed urethane to minimize heat exchange. May be used for short term storage of dry ice-solvent, liquid nitrogen, or for use as a hot bath. They are ideal for storing frozen biological specimens.
Height: 324 mm
I.D.: 146 mm