Learn More
Electron Microscopy Sciences Polyethylene Gloves, Large
SDP
Supplier:  Electron Microscopy Sciences 71096
Encompass_Preferred
Disposable low cost ambidextrous gloves provide protection to hands from chemical contamination. 1 3/4 mil thickness with excellent sensitivity. The gloves are powderless and sulfur free. Comes in a dispenser box.
Size: Large
Catalog No. 50-930-777