Learn More
Electron Microscopy Sciences Polyethylene Gloves, Small
SDP
Catalog No. 5027953
Encompass_Preferred
Disposable low cost ambidextrous gloves provide protection to hands from chemical contamination. 1 3/4 mil thickness with excellent sensitivity. The gloves are powderless and sulfur free. Comes in a dispenser box.
Size: Small
This item is not returnable. View return policy
Catalog No. 50-279-53 Supplier Electron Microscopy Sciences Supplier No. 71097S
Disposable low cost ambidextrous gloves provide protection to hands from chemical contamination. 1 3/4 mil thickness with excellent sensitivity. The gloves are powderless and sulfur free. Comes in a dispenser box.
Size: Small