Learn More
Medchemexpress LLC Polyethylene glycol 100000 | 25322-68-3 | 100,000 (average) | (C2H4O)nH2O | 50g
SDP
Catalog No. 5000269910
Encompass_Preferred
PEG100000 (Polyethylene glycol 100000) is a polymer compound with good biocompatibility and biodegradability and is widely used in drug controlled release systems and bioadhesive materials[1]
Catalog No. 50-002-69910 Supplier Medchemexpress LLC Supplier No. HYY0873A150G
Add to Cart
Add to Cart
PEG100000 (Polyethylene glycol 100000) is a polymer compound with good biocompatibility and biodegradability and is widely used in drug controlled release systems and bioadhesive materials[1]