Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Polyethylene Glycol 1450 NF (Flake)
SDP

Catalog No. 14186215 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
5 kg
10 kg
25 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-215 2.5 kg
14-186-104 100 g
14-186-037 5 kg
14-185-453 25 kg
14-185-497 10 kg
14-185-504 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-186-215 Supplier Medisca Supplier No. 077801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polyethylene Glycol 1450 NF flakes are white or almost white with a waxy or paraffin-like appearance. This Polyethylene Glycol has a molecular weight of 1450.

Polyethylene Glycol 1450 NF flakes are white or almost white with a waxy or paraffin-like appearance. This Polyethylene Glycol has a molecular weight of 1450. For use as an excipient.

Specifications

Chemical Name or Material Polyethylene Glycol 1450 NF (Flake)
Packaging Pail, 8 Liter, White, HDPE, Square
CAS 25322-68-3
Type Drug Standard
Molecular Weight (g/mol) 285 to 315 (average)
Quantity 2.5 kg
Percent Purity 97 to 103 %
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.