Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 1450, Waxy Soft Solid, BAKER™, J.T. Baker™

Catalog No. 02003637 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-637 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-637 Supplier Avantor Supplier No. U22007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Physical Form Waxy Soft Solid
Quantity 500 mL
Packaging Poly Bottle
Specific Gravity 1L = 1.15kg
Formula Weight 1300 to 1600 (Average)
Chemical Name or Material Polyethylene Glycol 1450
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.