Learn More
Medchemexpress LLC Polyethylene glycol 200 | 25322-68-3 | 98.0% | 200.00 g/mol | (C2H4O)nH2O | 25g
SDP
Catalog No. 5000268563
Encompass_Preferred
PEG200 (Polyethylene glycol 200) a neutral polymer of molecular weight 200 is a water-soluble low immunogenic and biocompatible polymer formed by repeating units of ethylene glycol[1]
Catalog No. 50-002-68563 Supplier Medchemexpress LLC Supplier No. HYY0873K25G
PEG200 (Polyethylene glycol 200) a neutral polymer of molecular weight 200 is a water-soluble low immunogenic and biocompatible polymer formed by repeating units of ethylene glycol[1]