Learn More
Medchemexpress LLC Polyethylene glycol 200 | 25322-68-3 | MFCD00081839 | 98.0% | 200.00 | (C2H4O)nH2O | 100g
SDP
Catalog No. 5000268739
Encompass_Preferred
PEG200 (Polyethylene glycol 200) a neutral polymer of molecular weight 200 is a water-soluble low immunogenic and biocompatible polymer formed by repeating units of ethylene glycol[1]
Catalog No. 50-002-68739 Supplier Medchemexpress LLC Supplier No. HYY0873K100G
PEG200 (Polyethylene glycol 200) a neutral polymer of molecular weight 200 is a water-soluble low immunogenic and biocompatible polymer formed by repeating units of ethylene glycol[1]