Learn More
Medchemexpress LLC Polyethylene glycol 2000 | 25322-68-3 | MFCD00081839 | 2000.00 | H(OCH2CH2)nOH | 100g
SDP
Catalog No. 5000268776
Encompass_Preferred
PEG2000 (Polyethylene glycol 2000) is a solvent for a large number of substances PEG2000 can be used as a carrier material and modifying agent PEG2000 is widely used in a variety of pharmaceutical formulations[1]
Catalog No. 50-002-68776 Supplier Medchemexpress LLC Supplier No. HYY0873B100G
PEG2000 (Polyethylene glycol 2000) is a solvent for a large number of substances PEG2000 can be used as a carrier material and modifying agent PEG2000 is widely used in a variety of pharmaceutical formulations[1]