Learn More
Medchemexpress LLC Polyethylene glycol | 25322-68-3 | 500 | (C2H4O)nH2O | 25g
SDP
Catalog No. 5000265915
Encompass_Preferred
PEG500 (Polyethylene glycol 500) is a versatile polymer that s used as a solvent dispersant lubricant and more in industries like pharmaceuticals and biotechnology and it can also modify liposomes PEG500 is commonly used as a solvent or carrier for drugs helping with capsule endoscopy to improve image quality and cecum completion rates[1][2]
Catalog No. 50-002-65915 Supplier Medchemexpress LLC Supplier No. HYY0873M25G
PEG500 (Polyethylene glycol 500) is a versatile polymer that s used as a solvent dispersant lubricant and more in industries like pharmaceuticals and biotechnology and it can also modify liposomes PEG500 is commonly used as a solvent or carrier for drugs helping with capsule endoscopy to improve image quality and cecum completion rates[1][2]