Learn More
Medchemexpress LLC Polyethylene glycol 35000 | 25322-68-3 | MFCD00081839 | 98.0% | 35,000 g/mol | H(OCH2CH2)nOH | 100g
SDP
Catalog No. 5000268782
Encompass_Preferred
PEG35000 is a polyethylene glycol with a molecular weight of 35 000 that can be used as a carrier material and modifier in drug delivery systems[1]
Catalog No. 50-002-68782 Supplier Medchemexpress LLC Supplier No. HYY0873O100G
PEG35000 is a polyethylene glycol with a molecular weight of 35 000 that can be used as a carrier material and modifier in drug delivery systems[1]