Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol 4,000

Catalog No. AAA161510I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA161510I 5000 g
AAA1615130 250 g
AAA161510B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA161510I Supplier Thermo Scientific Chemicals Supplier No. A161510I
  • $590.00
    $501.65 / Each of 1

    Save $88.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used in gene delivery of protoplast transfection and incubation. It is used in the preparation of extraction for measuring cytosolic phosphoenolpyruvate carboxykinase (PEPCK) from plant tissues. It is useful to mediate DNA transformation into protoplasts derived from rice leaf sheaths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 4000
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

pH 5 to 7
Melting Point 58°C to 62°C
Quantity 5000 g
Odor Odorless
Merck Index 14,7568
Solubility Information Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons.
Chemical Name or Material Polyethylene glycol 4,000
RTECSNumber TQ4050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.