Learn More
Polyethylene Glycol 40 Castor Oil, Hydrogenated, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Specifications
Specifications
| Color | Yellow liquid |
| Acid Value | 3 |
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Grade | Reagent |
| Hydroxyl Value | 59-68 |
| Packaging | Glass Bottle |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.