Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 400, MilliporeSigma™

Catalog No. MPX1286B2 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX1286B2 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX1286B2 Supplier MilliporeSigma Supplier No. PX1286B2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polyethylene Glycol 400; EMD Chemicals; 1L; Glass Bottle; F.W. HO(CH2CH2O)nH

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyethylene Glycol 400
CAS 25322-68-3
Quantity 1 L
Packaging Glass Bottle
Molecular Formula (C2H4O)n
Linear Formula HO(CH2CH2O)nH
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES [H]OCCO
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.07
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.