Learn More
Description
Polyethylene Glycol 400 NF is a clear to slightly hazy, colorless or practically colorless, viscous liquid. Has a slight characteristic odor. Slightly hygroscopic. This Polyethylene Glycol has a molecular weight of 400. For use as an excipient.
Specifications
Specifications
| Chemical Name or Material | Polyethylene Glycol 400 NF (Liquid) |
| Packaging | Jug, 4L, White, HDPE, Square, 38-400, TE Pano Neck |
| CAS | 25322-68-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 285 to 315 (average) |
| Quantity | 4 L |
| Percent Purity | 97 to 103 % |
| Grade | NF |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
