Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 400, TCI America™
SDP

Catalog No. N044325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N044325G 25 g
N0443500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N044325G Supplier TCI America Supplier No. N044325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG 400
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color Colorless
Physical Form Viscous Clear Liquid at 20°C
Quantity 25 g
Chemical Name or Material Polyethylene Glycol 400
EINECSNumber (7)-0129
RTECSNumber TQ3675000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.