Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol 600

Catalog No. p-3173400
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2179830 250 g
AAB217980B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2179830 Supplier Thermo Scientific Chemicals Supplier No. B2179830
Only null left
Add to Cart

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

Polyethylene glycol is used as a humectant in inks and adhesives. It acts as a solvent in soldering flux and as a dye carrier in inks. It is also useful as a modifier and binder in latex paint. It is used as an additive in synthetic lubricants, cutting oil and hydraulic fluids. It plays an important role for the precipitation of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Melting Point 17°C to 23°C
Density 1.12
Solubility Information Miscible with water,many organic solvents,aliphatic ketones,alcohols,chloroform,glycol ethers,esters,and aromatic hydrocarbons. Immiscible with ether and aliphatic hydrocarbons.
Quantity 250 g
Flash Point 252°C (485°F)
Refractive Index 1.468
Chemical Name or Material Polyethylene glycol 600
Hazard Category H227-H303-H315-H320-H333-H335
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
EINECSNumber 500-038-2
RTECSNumber TQ3800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.