Learn More
Medchemexpress LLC Polyethylene glycol 6000 | 25322-68-3 | MFCD00081839 | 99.9% | 6000.00 g/mol | H(OCH2CH2)nOH | 25g
SDP
Catalog No. 5000268737
Encompass_Preferred
PEG6000 can be used as an excipient such as Ointment base lubricant etc Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1]
Catalog No. 50-002-68737 Supplier Medchemexpress LLC Supplier No. HYY0873I25G
PEG6000 can be used as an excipient such as Ointment base lubricant etc Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1]