Learn More
Medchemexpress LLC Polyethylene glycol 8000 | 25322-68-3 | MFCD00081839 | 99.9% | Average molecular weight ≈8,000 g/mol | H(OCH2CH2)nOH | 100g
SDP
Catalog No. 5000268738
Encompass_Preferred
PEG8000 can be used for phage precipitation isolation of plasmid DNA and promotion of flat end-junction reactions[1]
Catalog No. 50-002-68738 Supplier Medchemexpress LLC Supplier No. HYY0873J100G
PEG8000 can be used for phage precipitation isolation of plasmid DNA and promotion of flat end-junction reactions[1]