Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 8000 (PEG 8000), Reagents

Catalog No. 01337129
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-129 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-129 Supplier Reagents Holdings Llc Supplier No. C22750003A5
Add to Cart
Add to Cart

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Polyethylene Glycol 8000 (PEG 8000), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 8000
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym Carbowax 8000, PEG 8000
SMILES [H]OCCO

Specifications

Color White
pH 4.5 to 7.5
Physical Form Solid
Boiling Point >200°C
Melting Point 60°C to 63°C
Avg. Mol. Wt. or Mol. Wt. Range 8000
Quantity 3 kg
Packaging Poly Bottle
Specific Gravity 1.0872
Chemical Name or Material Polyethylene Glycol 8000 (PEG 8000)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.