Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene glycol averag20KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 20239820KG
Encompass
Poly(ethylene glycol) (PEG) is a synthetic hydrophilic biocompatible polymer with widespread use in biomedical and other applications. PEGs are non-toxic fDA-approved generally non-immunogenic and frequently used in many biomedical applications including bioconjugation drug delivery surface functionalization and tissue engineering. In drug delivery PEGs can be used as linkers for antibody-drug conjugates (ADCs) or as a surface coating on nanoparticles to improve systemic drug delivery.
Catalog No. 50-290-2849