Learn More
Sigma Aldrich Fine Chemicals Biosciences Polyethylene glycol averag20KG
Catalog No. 502903561
Encompass
Polyethylene glycol (PEG) is a hydrophilic polymer. It can be easily synthesized by the anionic ring opening polymerization of ethylene oxide into a range molecular weights and variety of end groups. When crosslinked into networks PEG can have high water content forming hydrogels. Hydrogel formation can be initiated by either crosslinking PEG by ionizing radiation or by covalent crosslinking of PEG macromers with reactive chain ends. PEG is a suitable material for biological applications because it does not trigger an immune response.
Catalog No. 50-290-3561 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 20237120KG
Polyethylene glycol (PEG) is a hydrophilic polymer. It can be easily synthesized by the anionic ring opening polymerization of ethylene oxide into a range molecular weights and variety of end groups. When crosslinked into networks PEG can have high water content forming hydrogels. Hydrogel formation can be initiated by either crosslinking PEG by ionizing radiation or by covalent crosslinking of PEG macromers with reactive chain ends. PEG is a suitable material for biological applications because it does not trigger an immune response.